Anti-BRINP1 Rabbit Polyclonal Antibody (FITC (Fluorescein Isothiocyanate))
Catalog # BOSSBS-2761R-FITC
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:bone morphogenetic protein/retinoic acid inducible neural-specific 1
- Antigen symbol:BRINP1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:1620
- Antigen synonyms:DBCCR1|FAM5A|DBC1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:130-180/761
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibits cell proliferation by negative regulation of the G1/S transition. Mediates cell death which is not of the classical apoptotic type and regulates expression of components of the plasminogen pathway.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: BRINP1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat