Anti-ODC Rabbit Polyclonal Antibody (Alexa Fluor® 350)
BOSSBS-1294R-A350
: Bioss
- Antibody type:Primary
- Antigen name:Ornithine Decarboxylase
- Antigen symbol:ODC
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 350
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:89874
- Antigen synonyms:Ornithine Decarboxylase 1|ODC1|ornithine decarboxylase 1-like|ornithine decarboxylase antizyme inhibitor|AZI
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:320-370/461
- Purification:Purified by Protein A
- Pk:100 µl
Transports C5-C7 oxodicarboxylates across the inner membranes of mitochondria. Can transport 2-oxoadipate, 2-oxoglutarate, adipate, glutarate, and to a lesser extent, pimelate, 2-oxopimelate, 2-aminoadipate, oxaloacetate, and citrate.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: ODC1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat