Anti-GABRR1 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Catalog # BOSSBS-12084R-PE
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:Gamma-aminobutyric acid (GABA) A Receptor rho 1
- Antigen symbol:GABRR1
- Clonality:Polyclonal
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:2569
- Antigen synonyms:Gamma-aminobutyric acid receptor subunit rho-1|GABA(A) receptor subunit rho-1|GABA(C) receptor|GABRR1
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:210-260/479
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
GABA, the major inhibitory neurotransmitter in the vertebrate brain, mediates neuronal inhibition by binding to the GABA/benzodiazepine receptor and opening an integral chloride channel. Rho-1 GABA receptor could play a role in retinal neurotransmission.
Recommended Dilutions: Flow Cytometry: 1:20-100
Type: Primary
Antigen: GABRR1
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse