Specifications
- Antibody type:Primary
- Antigen name:Solute Carrier Family 12 Member A6
- Antigen symbol:SLC12A6
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Antigen synonyms:Solute carrier family 12|KCC3B|Potassium chloride cotransporter KCC3a S3|KCC3A|member 6|KCC 3|Gaxp|KCC3 A|KCC 3B|Solute carrier family 12 potassium/chloride transporters|KCC 3A|ACCPN|Potassium chloride cotransporter 3|S12A6_HUMAN.|KCC3|SLC12A 6|KCC3 B|Furosemide sensitive KCl cotransporter 3|SLC12 A6
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
The four isoforms of potassium/chloride co-transport channels (KCC) belong to a superfamily of cation-chloride co-transporters involved in cell volume maintenance. Nitric oxide (NO) donors activate KCCs, while inhibitors of the cGMP pathway prevent NO donor activation. The ubiquitously expressed KCC1 contains 12 transmembrane domains with both cytoplasmic N and C terminal domains. KCC2 expression is limited to neuronal tissues by a restrictive element similar to the neuronal-restrictive silencing factor. In neurons, KCC2 expression is correlated with an inhibitory response to GABA, while the absence of KCC2 is necessary for an unusual excitatory response to GABA. Alterations of KCC2 expression in the inferior colliculus of rat brain may be related to seizure susceptibility. Conversely, KCC3 is not suspected to play a major role in epilepsy. The two splice variants of KCC3, KCC3a and KCC3b, are predominantly expressed in brain and kidney, respectively, while KCC4 is expressed in muscle, brain, lung, heart and kidney.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: SLC12A6
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat