Anti-RFC2 Rabbit Polyclonal Antibody (Alexa Fluor® 555)
BOSSBS-11305R-A555
: Bioss
- Antibody type:Primary
- Antigen name:Replication Factor C subunit 2
- Antigen symbol:RFC2
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 555
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5982
- Antigen synonyms:Replication factor C 40 kDa subunit|Activator 1 subunit 2|RFC40|replication factor C subunit 2|Activator 1 40 kDa subunit
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:325-354/354
- Purification:Purified by Protein A
- Pk:100 µl
The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins proliferating cell nuclear antigen (PCNA) and activator 1. This subunit binds ATP (By similarity).
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RFC2
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 555
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat