- Antibody type:Primary
- Antigen name:Replication Factor C subunit 1
- Antigen symbol:RFC1
- Clonality:Polyclonal
- Conjugation:FITC (Fluorescein Isothiocyanate)
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Cross adsorption:No
- Form:liquid
- Gene ID:5981
- Antigen synonyms:Activator 1 subunit 1|Activator 1 140 kDa subunit|Replication factor C 140 kDa subunit|RF-C 140 kDa subunit|RFC1|Replication factor C large subunit|A1 140 kDa subunit|RFC140|DNA-binding protein PO-GA|Replication factor C subunit 1|Activator 1 large subunit
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:420-465/1148
- Purification:Purified by Protein A
- Pk:100 µl
The elongation of primed DNA templates by DNA polymerase delta and epsilon requires the action of the accessory proteins PCNA and activator 1. This subunit binds to the primer-template junction. Binds the PO-B transcription element as well as other GA rich DNA sequences. Could play a role in DNA transcription regulation as well as DNA replication and/or repair. Can bind single- or double-stranded DNA. Interacts with C-terminus of PCNA. 5' phosphate residue is required for binding of the N-terminal DNA-binding domain to duplex DNA, suggesting a role in recognition of non-primer template DNA structures during replication and/or repair.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: RFC1
Clonality: Polyclonal
Clone:
Conjugation: FITC (Fluorescein Isothiocyanate)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat