Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-GRK4, GRK5, GRK6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11235R-A350
 :  Bioss
Anti-GRK4, GRK5, GRK6 Rabbit Polyclonal Antibody (Alexa Fluor® 350)
  BOSSBS-11235R-A350
 :  Bioss

 

  • Antibody type:
    Primary
  • Antigen name:
    G Protein-coupled Receptor Kinase 4, 5, and 6
  • Antigen symbol:
    GRK4, GRK5, GRK6
  • Clonality:
    Polyclonal
  • Conjugation:
    Alexa Fluor® 350
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    2868, 2869, 2870
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    200-250/576
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

 

 

This gene encodes a member of the guanine nucleotide-binding protein (G protein)-coupled receptor kinase subfamily of the Ser/Thr protein kinase family. Such receptor phosphorylation initiates beta-arrestin-mediated receptor desensitization, internalization, and signaling events leading to their down-regulation.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: GRK4, GRK5, GRK6
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 350
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat