Anti-CD200R1 Rabbit Polyclonal Antibody (Alexa Fluor® 647)
Catalog # BOSSBS-10521R-A647
Supplier: Bioss
Specifications
- Antibody type:Primary
- Antigen name:CD200 Receptor 1
- Antigen symbol:CD200R1
- Clonality:Polyclonal
- Conjugation:Alexa Fluor® 647
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Gene ID:131450
- Antigen synonyms:MOX2R|HCRTR2|CD200R|OX2R
- Storage buffer:Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
- Storage temperature:Store at 4°C for 12 months
- Concentration:1 μg/μl
- Shipping temperature:4°C
- Immunogen:210-260/325
- Purification:Purified by Protein A
- Pk:100 µl
Specifications
About this item
Inhibitory receptor for the CD200/OX2 cell surface glycoprotein. Limits inflammation by inhibiting the expression of proinflammatory molecules including TNF-alpha, interferons, and inducible nitric oxide synthase (iNOS) in response to selected stimuli. Also binds to HHV-8 K14 viral CD200 homolog with identical affinity and kinetics as the host CD200.
Recommended Dilutions: IF(IHC-P): 1:50-200
Type: Primary
Antigen: CD200R1
Clonality: Polyclonal
Clone:
Conjugation: Alexa Fluor® 647
Epitope:
Host: Rabbit
Isotype: IgG1
Reactivity: Human