Order Entry
United Kingdom
ContactUsLinkComponent
Anti-VIP Rabbit Polyclonal Antibody (Cy5®)
Anti-VIP Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-0077R-CY5
Supplier:  Bioss
Anti-VIP Rabbit Polyclonal Antibody (Cy5®)
Catalog # BOSSBS-0077R-CY5
Supplier:  Bioss

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Vasoactive Intestinal Peptide
  • Antigen symbol:
    VIP
  • Clonality:
    Polyclonal
  • Conjugation:
    Cy5®
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Cross adsorption:
    No
  • Form:
    liquid
  • Gene ID:
    7432
  • Antigen synonyms:
    Pituitary adenylate cyclase-activating polypeptide type II receptorPACAP type II receptor|VIP-R-1|PACAP-R-2|PHM27
  • Storage buffer:
    Aqueous buffered solution containing 100ug/ml BSA, 50% glycerol and 0.09% sodium azide. Store at 4°C for 12 months.
  • Storage temperature:
    Store at 4°C for 12 months
  • Concentration:
    1 μg/μl
  • Shipping temperature:
    4°C
  • Immunogen:
    125-152/170
  • Purification:
    Purified by Protein A
  • Pk:
    100 µl

Specifications

About this item

VIP causes vasodilation, lowers arterial blood pressure, stimulates myocardial contractility, increases glycogenolysis and relaxes the smooth muscle of trachea, stomach and gall bladder. PHM and PHV also cause vasodilation. PHM-27 is a potent agonist of the calcitonin receptor CALCR, with similar efficacy as calcitonin.

Recommended Dilutions: IF(IHC-P): 1:50-200

Type: Primary
Antigen: VIP
Clonality: Polyclonal
Clone:
Conjugation: Cy5®
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat