HPLC columns, YMC-Pack™ YMCbasic
Product Details & Documents
The proprietary derivatisation procedure for YMCbasic produces a material with controlled surface coverage, which gives excellent lot-to-lot reproducibility. Used as an alternative bonding approach to reduce peak tailing of basic pharmaceuticals.
- No need for ion pair reagents or amine modifiers
- Complementing selectivity to C8 and C18 materials
- High resolution and excellent peak symmetry for basic compound
YMCbasic is suitable for separation of acidic compounds, showing slight retention of highly polar acid compounds such as maleate. This provides a complementing selectivity with conventional C8 and C18 materials, but without peak tailing for basic compounds.
: Every column supplied with lot certificate on physicochemical properties of bare silica, minimal metal traces, bonding and chromatographic performance; test chromatogram showing high performance plate count together with tight specification on peak symmetry.
Specifications
- Column Format:Analytical
- Ø int.:4,60 mm
- Particle Size:3 µm
- Pore size:200 Å
- Length:250 mm
- Phase:YMCbasic
- Carbon load:7%
- Cat. No.:YMCABA99S032546WT
- Endcapped:Yes
- USP code:L7
- pH range:2,0 - 7,5
- Surface area:175 m²/g
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



