Anti-CASP5 p20 Rabbit Polyclonal Antibody [clone: D121]
Catalog # BWRLBS7048
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CASP5 p20
- Clonality:Polyclonal
- Clone:D121
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 100-150 of Human Caspase-5.
Caspase-5 p20 (D121) pAb detects endogenous levels of the large subunit of active caspase-5 resulting from cleavage at Asp121.
Type: Primary
Antigen: CASP5 p20
Clonality: Polyclonal
Clone: D121
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat