Anti-Ac-Histone H3 Rabbit Polyclonal Antibody [clone: K18]
Catalog # BWRLBS70006
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Ac-Histone H3
- Clonality:Polyclonal
- Clone:K18
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide including acetyl-lysine 18 of Histone H3.
Ac-Histone H3 (K18) pAb detects endogenous levels of Histone H3 protein only when acetylated at Lys18.
Type: Primary
Antigen: Ac-Histone H3
Clonality: Polyclonal
Clone: K18
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat