Anti-BrdU Mouse Monoclonal Antibody
BWRLMB6004
- Antibody type:Primary
- Antigen symbol:BrdU
- Clonality:Monoclonal
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Multiple
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Recombinant full length Human BrdU.
BrdU Mab detects endogenous levels of BrdU protein.
Type: Primary
Antigen: BrdU
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype:
Reactivity: All