Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl R8]
Catalog # BWRLBS7512
Anti-Histone H3 Rabbit Polyclonal Antibody [clone: Mono-Methyl R8]
Catalog # BWRLBS7512

Specifications

  • Antibody type:
    Primary
  • Antigen symbol:
    Histone H3
  • Clonality:
    Polyclonal
  • Clone:
    Mono-Methyl R8
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Size:
    100 µg
  • Pk:
    100 µG

Specifications

About this item

Synthetic peptide conjugated to KLH derived from within residues 1-100 of Human Histone H3, Mono-methylated at R8.

Histone H3 (Mono-Methyl R8) pAb detects endogenous levels of histone H3 only when Mono-Methyled at Arg8.

Type: Primary
Antigen: Histone H3
Clonality: Polyclonal
Clone: Mono-Methyl R8
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat; Other (Wide Range)