Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-p-Rpb1 Rabbit Polyclonal Antibody [clone: S1619]
  BWRLBS4246
Anti-p-Rpb1 Rabbit Polyclonal Antibody [clone: S1619]
  BWRLBS4246

 

  • Antibody type:
    Primary
  • Antigen symbol:
    p-Rpb1
  • Clonality:
    Polyclonal
  • Clone:
    S1619
  • Host:
    Rabbit
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Synthetic phosphopeptide derived from human Rpb1 around the phosphorylation site of Serine 1619.

p-Rpb1 (S1619) pAb detects endogenous levels of Rpb1 protein only when phosphorylated at Ser1619.

Type: Primary
Antigen: p-Rpb1
Clonality: Polyclonal
Clone: S1619
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat