Anti-p-SMC 1 Rabbit Polyclonal Antibody [clone: S957]
BWRLBS4174
- Antibody type:Primary
- Antigen symbol:p-SMC 1
- Clonality:Polyclonal
- Clone:S957
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human SMC 1 around the phosphorylation site of Serine 957.
p-SMC1 (S957) pAb detects endogenous levels of SMC1protein only when phosphorylated at Ser957
Type: Primary
Antigen: p-SMC 1
Clonality: Polyclonal
Clone: S957
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse