Anti-p-NMDAzeta1 Rabbit Polyclonal Antibody [clone: S897]
Catalog # BWRLBS4801
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p-NMDA zeta1
- Clonality:Polyclonal
- Clone:S897
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic phosphopeptide derived from human NMDAzeta1 around the phosphorylation site of Serine 897.
p-NMDA?1 (S897) pAb detects endogenous levels of NMDA?1 protein only when phosphorylated at Ser897.
Type: Primary
Antigen: p-NMDAzeta1
Clonality: Polyclonal
Clone: S897
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat