Anti-p-Tau Rabbit Polyclonal Antibody [clone: T212]
BWRLBS4775
- Antibody type:Primary
- Antigen symbol:p-Tau
- Clonality:Polyclonal
- Clone:T212
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Synthetic phosphopeptide derived from human isoforms Tau-4 around the phosphorylation site of Threonine 212.
p-Tau (T212) pAb detects endogenous levels of Tau only when phosphorylated at Thr529, isoforms Tau-3 and isoforms Tau-4 phosphorylated at Thr212, and other isoforms phosphorylated at the corresponding Site.
Type: Primary
Antigen: p-Tau
Clonality: Polyclonal
Clone: T212
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat