Anti-PIG11 Rabbit Polyclonal Antibody [clone: P93]
Catalog # BWRLBS1865
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:PIG11
- Clonality:Polyclonal
- Clone:P93
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 71-120 of Human PIG11.
PIG11 (P93) pAb detects endogenous levels of PIG11 protein.
Type: Primary
Antigen: PIG11
Clonality: Polyclonal
Clone: P93
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human