Anti-NRF-1 Rabbit Polyclonal Antibody [clone: V256]
BWRLBS3233
- Antibody type:Primary
- Antigen symbol:NRF-1
- Clonality:Polyclonal
- Clone:V256
- Host:Rabbit
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 224-276 of Human NRF-1.
NRF-1 (V256) pAb detects endogenous levels of NRF-1 protein.
Type: Primary
Antigen: NRF-1
Clonality: Polyclonal
Clone: V256
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat