Anti-UTP14A Rabbit Polyclonal Antibody [clone: E30]
Catalog # BWRLBS2671
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:UTP14A
- Clonality:Polyclonal
- Clone:E30
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to the N-terminual of Human UTP14A.
UTP14A (E30) pAb detects endogenous levels of UTP14A protein.
Type: Primary
Antigen: UTP14A
Clonality: Polyclonal
Clone: E30
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse