Anti-ERF Rabbit Polyclonal Antibody [clone: E520]
BWRLBS1744
- Antibody type:Primary
- Antigen symbol:ERF
- Clonality:Polyclonal
- Clone:E520
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 500-550 of Human ERF.
ERF (E520) pAb detects endogenous levels of ERF protein.
Type: Primary
Antigen: ERF
Clonality: Polyclonal
Clone: E520
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse