Anti-CysLTR2 Rabbit Polyclonal Antibody [clone: K322]
Catalog # BWRLBS2614
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:CysLTR2
- Clonality:Polyclonal
- Clone:K322
- Host:Rabbit
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 290-336 of Human CysLTR2.
CysLTR2 (K322) pAb detects endogenous levels of CysLTR2 protein.
Type: Primary
Antigen: CysLTR2
Clonality: Polyclonal
Clone: K322
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat