Anti-Cytochrome b5 Rabbit Polyclonal Antibody [clone: G82]
BWRLBS2410
- Antibody type:Primary
- Antigen symbol:Cytochrome b5
- Clonality:Polyclonal
- Clone:G82
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 51-100 of Human Cytochrome b5.
Cytochrome b5 (G82) pAb detects endogenous levels of Cytochrome b5protein.
Type: Primary
Antigen: Cytochrome b5
Clonality: Polyclonal
Clone: G82
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat