Anti-Rhodopsin Rabbit Polyclonal Antibody [clone: L328]
BWRLBS1591
- Antibody type:Primary
- Antigen symbol:Rhodopsin
- Clonality:Polyclonal
- Clone:L328
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 301-350 of Human Rhodopsin.
Rhodopsin (L328) pAb detects endogenous levels of Rhodopsin protein.
Type: Primary
Antigen: Rhodopsin
Clonality: Polyclonal
Clone: L328
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat