Anti-ERK 2 Rabbit Polyclonal Antibody [clone: R172]
BWRLBS2265
- Antibody type:Primary
- Antigen symbol:ERK 2
- Clonality:Polyclonal
- Clone:R172
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 140-190 of Human ERK 2.
ERK 2 (R172) pAb detects endogenous levels of ERK 2 protein.
Type: Primary
Antigen: ERK 2
Clonality: Polyclonal
Clone: R172
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat