Anti-Calregulin Rabbit Polyclonal Antibody [clone: G54]
Catalog # BWRLBS2264
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Calregulin
- Clonality:Polyclonal
- Clone:G54
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 20-70 of Human Calregulin.
Calregulin (G54) pAb detects endogenous levels of Calregulin protein
Type: Primary
Antigen: Calregulin
Clonality: Polyclonal
Clone: G54
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat