Anti-Integrin beta3 Rabbit Polyclonal Antibody [clone: S778]
Catalog # BWRLBS3660
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Integrin beta3
- Clonality:Polyclonal
- Clone:S778
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 751-800 of Human Integrin beta3.
Integrin ?3 (S778) pAb detects endogenous levels of Integrin ?3 protein.
Type: Primary
Antigen: Integrin beta3
Clonality: Polyclonal
Clone: S778
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat