Anti-Septin 7 Rabbit Polyclonal Antibody [clone: M368]
Catalog # BWRLBS3937
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:Septin 7
- Clonality:Polyclonal
- Clone:M368
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 332-389 of Human Septin 7.
Septin 7 (M368) pAb detects endogenous levels of Septin 7 protein.
Type: Primary
Antigen: Septin 7
Clonality: Polyclonal
Clone: M368
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat