Anti-Neuregulin-1/SMDF Rabbit Polyclonal Antibody [clone: Y4]
BWRLBS1173
- Antibody type:Primary
- Antigen symbol:Neuregulin-1/SMDF
- Clonality:Polyclonal
- Clone:Y4
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to the N-terminal of Human Neuregulin-1/SMDF.
SMDF/Neuregulin 1 (Y4) pAb detects endogenous levels of SMDF/Neuregulin 1 protein.
Type: Primary
Antigen: Neuregulin-1/SMDF
Clonality: Polyclonal
Clone: Y4
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat