Anti-Ras Rabbit Polyclonal Antibody [clone: H27]
BWRLBS1309
- Antibody type:Primary
- Antigen symbol:Ras
- Clonality:Polyclonal
- Clone:H27
- Host:Rabbit
- ImmunoChemistry:Yes
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Size:100 µg
- Pk:100 µG
Synthetic peptide, corresponding to amino acids 1-50 of Human Ras.
Ras (H27) pAb detects endogenous levels of total K-Ras, H-Ras and N-Ras proteins.
Type: Primary
Antigen: Ras
Clonality: Polyclonal
Clone: H27
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human;Mouse;Rat