Anti-p14 ARF/p19 ARF Rabbit Polyclonal Antibody [clone: Q99]
Catalog # BWRLBS1266
Supplier: Bioworld Technology
Specifications
- Antibody type:Primary
- Antigen symbol:p14 ARF/p19 ARF
- Clonality:Polyclonal
- Clone:Q99
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Synthetic peptide, corresponding to amino acids 65-121 of Human p14 ARF/p19 ARF.
p14 ARF/p19 ARF (Q99) pAb detects endogenous levels of p14 ARF/p19 ARF protein.
Type: Primary
Antigen: p14 ARF/p19 ARF
Clonality: Polyclonal
Clone: Q99
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human