Order Entry
United Kingdom
ContactUsLinkComponent
Copper (II) ethylenediamine 1.0 M in water, Sigma-Aldrich®
Corrosive
Copper (II) ethylenediamine 1.0 M in water, Sigma-Aldrich®
Catalog # SIAL442305-1L
Supplier:  Merck
CAS Number:  
Corrosive
Copper (II) ethylenediamine 1.0 M in water, Sigma-Aldrich®
Catalog # SIAL442305-1L
Supplier:  Merck
CAS Number:  

Specifications

  • Pack type:
    Glass bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

Specification Test Results

  • Colour:
    Dark Blue
  • Form:
    Liquid
  • Infrared Spectrum:
    Conforms to Structure
  • Purity titration by NATH:
    0,95 - 1,10 M
  • Concentration:
    1,0 M in H₂O

Specification Test Results

About this item

 :
 :217.76 g/mol
Density:1.086 g/ml (25 °C)
ADR:8 (6.1),II
 :M-Clarity™ quality level = MQ200
UN:1761
 :GHS05
CAS Number:14552-35-3
Formula:Cu(H₂NCH₂CH₂NH₂)₂(OH)₂
MDL Number:MFCD00274628