Order Entry
United Kingdom
ContactUsLinkComponent
Tritirachium album proteinase K, MP Biomedicals
Tritirachium album proteinase K, MP Biomedicals
Supplier:  MP Biomedicals
Certificates
Tritirachium album proteinase K, MP Biomedicals
Supplier:  MP Biomedicals

Specifications

  • CAS No.:
    39450-01-6
  • Species:
    Tritirachium album
  • Storage conditions:
    +4 °C
  • Form:
    White/Off-white lyophilized powder
  • Enzyme name:
    Proteinase K
  • Enzyme activity:
    >30 mAnson u/mg
  • Molecular weight:
    ~30 kDa
  • pH:
    8,9

Specifications

Frequently Bought Together