Membrane filtration apparatus
Supplier: Restek
Certificates
About this item
This filtration apparatus are with fritted glass support that is recommended for routine filtration of corrosive liquids and removal of particles from LC solvents. The ground joint connection eliminates phthalate contamination that can occur when using silicone or neoprene stoppers. The support base has a coarse-porosity glass frit and an integral vacuum connection, located above the drip tip to prevent contamination of the vacuum line with filtrate droplets.
Delivery: Apparatus includes a funnel, an anodized aluminum clamp, a 47 mm fritted glass support base and a filtration flask.
Frequently Bought Together