Anti-SIGLEC8 Rabbit Polyclonal Antibody
ABNOPAB30457
: Abnova
- Antibody type:Primary
- Antigen name:Sialic Acid Binding Ig Like Lectin 8 Recombinant
- Antigen symbol:SIGLEC8
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Form:Liquid
- Gene ID:27181
- Storage buffer:In PBS, pH 7.2 (40% glycerol, 0.02% sodium azide).
- Sequence:RSCRKKSARPAAGVGDTGMEDAKAIRGSASQGPLTESWKDGNPLKKPPPAVAPSSGEEGELHYATLSFHKVKPQDPQGQEATDSEYSEIKIHKRETAETQACLRNHNPSSKEVRG
- Storage temperature:Store at 4° C. For long term storage store at -20° C.
- Immunogen:Recombinant SIGLEC8.
- Purification:Antigen affinity purification
- Size:100 µL
- Pk:100 µl
Rabbit polyclonal antibody raised against partial recombinant human SIGLEC8.
Type: Primary
Antigen: SIGLEC8
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human