High purity airgas VOC regulators
Supplier: Restek
Certificates
About this item
These single stage stainless steel VOC regulators features stainless steel diaphragm, two pressure gauges, CGA-180 fitting and Kel-F® seat.
- Accurate pressure control even at low flow rates
- Individually tested for leaks and impurities
Specifications
- Catalog No:
- REST26371
- REST26372
- Type:
- Single stage
- Single stage
- Max. inlet pressure:
- 206,8 bar
- 206,8 bar
- Port:
- 1/4" FNTP
- 1/4" FNTP