Capillary columns, MXT®-65 (Siltek®-treated stainless steel)
Supplier: Restek
Certificates
About this item
MXT®-65 columns are general-purpose columns for phenols, triglycerides, and fatty acids.
- Mid to high polarity phase
- Crossbond® diphenyl dimethyl polysiloxane
- Temperature range: 50 to 300 °C
- Equivalent to USP G17 phase
Frequently Bought Together