Order Entry
United Kingdom
ContactUsLinkComponent
Twin-column adsorption apparatus
Twin-column adsorption apparatus
Catalog # STAN14060-0
Supplier:  STANHOPE SETA
Twin-column adsorption apparatus
Catalog # STAN14060-0
Supplier:  STANHOPE SETA
Specifications
  • Description:
    Twin-column adsorption apparatus
  • Pk:
    1
Specifications
About this item
The twin-column adsorption apparatus can measures fluorescent indicator adsorption in liquid petroleum products.
  • Determines volume % of saturates, olefins and aromatics in petroleum fractions
  • Analyses two samples simultaneously
  • Integral ultra violet illumination