Tubing bender
Catalog # REST23009
Supplier: Restek
Specifications
- Description:Tubing bender
- Pk:1
Specifications
About this item
These benders assists in making accurate left-hand, right-hand or offset bends. Bends 3,17 mm, 4,76 mm, or 6,35 mm tubing.