Sterilisation wraps, Bio-Shield®
These versatile non woven wraps offer fluid repellency and provide an excellent barrier against airborne bacteria. The soft, drapable non woven blend of cellulose and polyester is virtually lint-free and helps protect wrapped contents from contamination.
- Wraps feature sterilisation tapes that adhere firmly and remove easily
- Superior bacterial barrier properties compared to linen
- Repels fluids
Frequently Bought Together