Anti-HMGB1 Mouse Monoclonal Antibody [clone: KS1]
Catalog # ENZOADICSA614E
Supplier: Enzo Life Sciences
Specifications
- Antibody type:Primary
- Antigen name:High-mobility Group Box 1
- Antigen symbol:HMGB1
- Clonality:Monoclonal
- Clone:KS1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Seq:02
- Form:Liquid
- Antigen synonyms:high-mobility group box 1|RP11-550P23.1|high mobility group protein 1|Amphoterin|SBP-1|high-mobility group (nonhistone chromosomal) protein 1|high mobility group protein B1|HMG3|Sulfoglucuronyl carbohydrate binding protein|HMG1|HMG-1
- Storage buffer:PBS containing 50% glycerol
- Storage temperature:–20 °C
- Shipping temperature:Shipped on blue ice
- Immunogen:Pig HMGB1 (HMG1)
- Purification:Purified by Protein A affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
The chromatin non-histone high mobility group of proteins (HMG)-1 and -2 contain conserved domains known as HMG-boxes. Both HMG-1 and HMG-2 contain an N-terminal HMG box, a central HMG box and an acidic carboxy terminus.
- Dilution Range: 5 µg/ml for Western blot
- Detects a band of ~29kDa by Western blot
HMG-1/2 have been shown to facilitate the binding of various sequence-specific transcription factors to their respective DNA binding sites, such as steroid hormone receptors, and p53.
Type: Primary
Antigen: High-mobility Group Box 1
Clonality: Monoclonal
Clone: KS1
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human, Mouse, Rat