Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD.3]
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD.3]
  BNUM0885-50
 :  Biotium
Anti-BrdU Mouse Monoclonal Antibody (Purified, BSA-free) [clone: BRD.3]
  BNUM0885-50
 :  Biotium

 

  • Antibody type:
    Primary
  • Antigen name:
    BrdU
  • Clonality:
    Monoclonal
  • Clone:
    Clone BRD.3
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Multiple
  • Antigen synonyms:
    BUdr|Bromodeoxyuridine
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

 

 

This antibody reacts with Bromodeoxyuridine (BrdU) in single stranded DNA (produced by partial denaturation of double stranded DNA), BrdU coupled to a protein carrier, as well as free BrdU. BrdU is a thymidine analog, incorporated into cell nuclei during DNA synthesis prior to mitosis. Antibody to BrdU is helpful in detecting S-phase cells, providing useful information on the aggressiveness of tumors.

Type: Primary
Antigen: Not Applicable
Clonality: Monoclonal
Clone: BRD.3
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: All species