Order Entry
United Kingdom
ContactUsLinkComponent
Anti-NAPSA Mouse Monoclonal Antibody (CF568) [clone: NAPSA/1239]
Anti-NAPSA Mouse Monoclonal Antibody (CF568) [clone: NAPSA/1239]
Supplier:  Biotium
Certificates
Anti-NAPSA Mouse Monoclonal Antibody (CF568) [clone: NAPSA/1239]
Supplier:  Biotium

Specifications

  • Catalog No:
  • BNC681239-500
  • BNC681239-100
  • Antigen symbol:
  • NAPSA
  • NAPSA
  • Antigen name:
  • Napsin A
  • Napsin A
  • Antigen synonyms:
  • NAPA|Aspartyl protease 4|Kidney derived aspartic protease like protein (Kdap)|KAP|Pronapsin A|NAP1|Napsa|SNAPA|napsin A aspartic peptidase|ASP4
  • NAPA|Aspartyl protease 4|Kidney derived aspartic protease like protein (Kdap)|KAP|Pronapsin A|NAP1|Napsa|SNAPA|napsin A aspartic peptidase|ASP4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone NAPSA/1239
  • Clone NAPSA/1239
  • Reactivity:
  • Human
  • Human
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 512843
  • 512843
  • Isotype:
  • IgG kappa
  • IgG kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

Specifications

Frequently Bought Together