Order Entry
United Kingdom
ContactUsLinkComponent
Anti-BCL2 Mouse Monoclonal Antibody [clone: 100/D5 124]
Anti-BCL2 Mouse Monoclonal Antibody [clone: 100/D5 124]
Catalog # BNUM1234-50
Supplier:  Biotium
Anti-BCL2 Mouse Monoclonal Antibody [clone: 100/D5 124]
Catalog # BNUM1234-50
Supplier:  Biotium

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    bcl-2
  • Antigen symbol:
    BCL2
  • Clonality:
    Monoclonal
  • Clone:
    Clone 100/D5 124
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Gene ID:
    150749
  • Antigen synonyms:
    Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
  • Storage buffer:
    PBS/0,05% azide
  • Concentration:
    1 mg/mL
  • Purification:
    BSA-free
  • Size:
    50 μL
  • Pk:
    50 µl

Specifications

About this item

This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.

  • Purified
  • BSA-free

Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 100/D5 124
Conjugation: Purified, BSA-free
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human