This antibody recognizes a protein of 25-26 kDa, identified as the bcl-2 α oncoprotein. It shows no cross-reaction with Bcl-x or Bax protein. Expression of bcl-2 α oncoprotein inhibits programmed cell death (apoptosis). In most follicular lymphomas, neoplastic germinal centers express high levels of bcl-2 α protein, whereas the normal or hyperplastic germinal centers are negative. Consequently, this antibody is valuable when distinguishing between reactive and neoplastic follicular proliferation in lymph node biopsies. It may also be used in distinguishing between those follicular lymphomas that express bcl-2 protein and the small number in which the neoplastic cells are bcl-2 negative.
Type: Primary
Antigen: BCL2
Clonality: Monoclonal
Clone: 100/D5 124
Conjugation: CF568
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human
- Catalog No:
- BNC681234-500
- BNC681234-100
- Antigen symbol:
- BCL2
- BCL2
- Antigen name:
- bcl-2
- bcl-2
- Antigen synonyms:
- Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
- Apoptosis regulator Bcl-2|B-cell CLL/lymphoma-2
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 568
- CF® 568
- Clone:
- Clone 100/D5 124
- Clone 100/D5 124
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Gene ID:
- 150749
- 150749
- Isotype:
- IgG
- IgG
- Western blot:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide