Anti-MSN Mouse Monoclonal Antibody [clone: MSN/493]
Catalog # ABNOMAB14363
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:MSN Recombinant
- Antigen symbol:MSN
- Clonality:Monoclonal
- Clone:MSN/493
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Environmentally Preferable:
- Form:Liquid
- Gene ID:4478
- Amino acid number:P26038
- Storage buffer:In 10 mM PBS (0.05% BSA, 0.05% sodium azide).
- Immunogen:Recombinant protein corresponding to full length human MSN.
- Purification:Protein A/G purification
- Size:100 µG
- Pk:100 µG
Specifications
About this item
Mouse monoclonal antibody raised against full length recombinant human MSN.
Type: Primary
Antigen: MSN
Clonality: Monoclonal
Clone: MSN/493
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1, kappa
Reactivity: Human