About this item
Myogenin is a member of the MyoD family of myogenic basic helix-loop-helix (bHLH) transcription factors that also includes MyoD, Myf-5, and MRF4 (also known as herculinor Myf-6). MyoD family members are expressed exclusively in skeletal muscle and play a key role in activating myogenesis by binding to enhancer sequences of muscle-specific genes. The regulatory domain of MyoD is approximately 70 amino acids in length and includes both a basic DNA binding motif and a bHLH dimerization motif. MyoD family members share about 80% amino acid homology in their bHLH motifs. Anti-myogenin labels the nuclei of myoblasts in developing muscle tissue, and is expressed in tumor cell nuclei of rhabdomyosarcoma and some leiomyosarcomas. Positive nuclear staining may occur in Wilms' tumor.
Type: Primary
Antigen: MYOG
Clonality: Monoclonal
Clone: MGN185 F5D
Conjugation: CF640R
Epitope:
Host: Mouse
Isotype: IgGs
Reactivity: Cat, Human, Mouse, Pig, Rat
Specifications
- Catalog No:
- BNC401233-100
- BNC401233-500
- Antigen symbol:
- MYOG
- MYOG
- Antigen name:
- Myogenin
- Myogenin
- Antigen synonyms:
- Class C basic helix-loop-helix protein 3|Myogenic factor 4|MYF4|bHLHc3|MYOG|cb553|Myf-4
- Class C basic helix-loop-helix protein 3|Myogenic factor 4|MYF4|bHLHc3|MYOG|cb553|Myf-4
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- CF® 640R
- CF® 640R
- Clone:
- Clone MGN185 F5D
- Clone MGN185 F5D
- Reactivity:
- Pig, Human, Rat, Mouse, Cat
- Pig, Human, Rat, Mouse, Cat
- Host:
- Mouse
- Mouse
- Gene ID:
- 2830
- 2830
- Isotype:
- IgGs
- IgGs
- ImmunoChemistry:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Concentration:
- 0,1 mg/ml
- 0,1 mg/ml
- Storage buffer:
- PBS/0,1% BSA/0,05% azide
- PBS/0,1% BSA/0,05% azide