Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-MYOG Mouse Monoclonal Antibody (CF488A) [clone: MGN185 F5D]
 :  Biotium
 
Anti-MYOG Mouse Monoclonal Antibody (CF488A) [clone: MGN185 F5D]
 :  Biotium

 

  • Catalog No:
  • BNC881233-100
  • BNC881233-500
  • Antigen symbol:
  • MYOG
  • MYOG
  • Antigen name:
  • Myogenin
  • Myogenin
  • Antigen synonyms:
  • Class C basic helix-loop-helix protein 3|Myogenic factor 4|MYF4|bHLHc3|MYOG|cb553|Myf-4
  • Class C basic helix-loop-helix protein 3|Myogenic factor 4|MYF4|bHLHc3|MYOG|cb553|Myf-4
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 488A
  • CF® 488A
  • Clone:
  • Clone MGN185 F5D
  • Clone MGN185 F5D
  • Reactivity:
  • Pig, Human, Rat, Mouse, Cat
  • Pig, Human, Rat, Mouse, Cat
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 2830
  • 2830
  • Isotype:
  • IgGs
  • IgGs
  • ImmunoChemistry:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together