Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-UACA Mouse Monoclonal Antibody (CF568) [clone: UACA/1222]
 :  Biotium
 
Anti-UACA Mouse Monoclonal Antibody (CF568) [clone: UACA/1222]
 :  Biotium

 

  • Catalog No:
  • BNC681222-500
  • BNC681222-100
  • Antigen symbol:
  • UACA
  • UACA
  • Antigen name:
  • UACA / Nucling
  • UACA / Nucling
  • Antigen synonyms:
  • Uveal autoantigen with coiled-coil domains and ankyrin repeats (UACA)|NUCLING|Nuclear membrane binding protein
  • Uveal autoantigen with coiled-coil domains and ankyrin repeats (UACA)|NUCLING|Nuclear membrane binding protein
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • CF® 568
  • CF® 568
  • Clone:
  • Clone UACA/1222
  • Clone UACA/1222
  • Reactivity:
  • Human, Mouse
  • Human, Mouse
  • Host:
  • Mouse
  • Mouse
  • Gene ID:
  • 108049
  • 108049
  • Isotype:
  • IgG1 kappa
  • IgG1 kappa
  • Western blot:
  • Yes
  • Yes
  • ImmunoFluorescence:
  • Yes
  • Yes
  • Concentration:
  • 0,1 mg/ml
  • 0,1 mg/ml
  • Storage buffer:
  • PBS/0,1% BSA/0,05% azide
  • PBS/0,1% BSA/0,05% azide

 

Frequently Bought Together